Assay Details
| Gene Symbol: NSMCE4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148346.2 | XP_015003832.1 | 1068 | 82 | ||
| GenBank mRNA | JU322698.1 | - | 963 | 82 | ||
| JU473691.1 | - | 1009 | 82 | |||
| JV043476.1 | - | 994 | 82 | |||
| JV717508.1 | - | 932 | 82 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NSMCE4A
Genomic Map