Assay Details
| Gene Symbol: SFT2D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265679.1 | NP_001252608.1 | 258 | 81 | ||
| XM_028833311.1 | XP_028689144.1 | 603 | 81 | |||
| XM_028833347.1 | XP_028689180.1 | 607 | 81 | |||
| GenBank mRNA | JV635607.1 | - | 258 | 81 | ||
More Information
Set Membership: |
|

SFT2D2
Genomic Map