Assay Details
| Gene Symbol: SV2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105846.4 | XP_001105846.1 | 824 | 66 | ||
| XM_015140296.2 | XP_014995782.1 | 824 | 66 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SV2C| Gene Symbol: SV2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105846.4 | XP_001105846.1 | 824 | 66 | ||
| XM_015140296.2 | XP_014995782.1 | 824 | 66 | |||
Set Membership: |
|