Assay Details
| Gene Symbol: LOC715257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU326862.1 | - | 1047 | 112 | ||
| JU476529.1 | - | 1172 | 112 | |||
| JV635566.1 | - | 1706 | 112 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC715257| Gene Symbol: LOC715257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU326862.1 | - | 1047 | 112 | ||
| JU476529.1 | - | 1172 | 112 | |||
| JV635566.1 | - | 1706 | 112 | |||
Set Membership: |
|