Assay Details
| Gene Symbol: PIGV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001109737.4 | XP_001109737.1 | 328 | 59 | ||
| XM_015132140.2 | XP_014987626.1 | 537 | 59 | |||
| XM_015132149.2 | XP_014987635.1 | 268 | 59 | |||
| XM_015132157.2 | XP_014987643.1 | 758 | 59 | |||
| XM_015132161.2 | XP_014987647.1 | 1215 | 59 | |||
| XM_015132170.2 | XP_014987656.1 | 1032 | 59 | |||
| XM_015132179.2 | XP_014987665.1 | 419 | 59 | |||
| XM_028837504.1 | XP_028693337.1 | 972 | 59 | |||
| XM_028837506.1 | XP_028693339.1 | 512 | 59 | |||
| GenBank mRNA | JU322912.1 | - | 131 | 59 | ||
| JU472639.1 | - | 146 | 59 | |||
| JV043498.1 | - | 89 | 59 | |||
| JV639321.1 | - | 204 | 59 | |||
More Information
Set Membership: |
|

PIGV
Genomic Map