Assay Details
| Gene Symbol: CDK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261307.1 | NP_001248236.1 | 726 | 64 | ||
| XM_015134230.2 | XP_014989716.1 | 1386 | 64 | |||
| XM_015134231.2 | XP_014989717.1 | 870 | 64 | |||
| XM_028844866.1 | XP_028700699.1 | 1456 | 64 | |||
| XM_028844867.1 | XP_028700700.1 | 1589 | 64 | |||
| GenBank mRNA | JU319885.1 | - | 592 | 64 | ||
| JU319886.1 | - | 592 | 64 | |||
| JU471047.1 | - | 726 | 64 | |||
| JU471048.1 | - | 726 | 64 | |||
More Information
Set Membership: |
|

CDK6
Genomic Map