Assay Details
| Gene Symbol: HDDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194872.3 | NP_001181801.1 | 293 | 75 | ||
| XM_002803907.2 | XP_002803953.1 | 574 | 75 | |||
| XM_028846840.1 | XP_028702673.1 | 315 | 75 | |||
| GenBank mRNA | JU322498.1 | - | 419 | 75 | ||
| JU475800.1 | - | 449 | 75 | |||
| JV047229.1 | - | 485 | 75 | |||
| JV668174.1 | - | 539 | 75 | |||
| JV716223.1 | - | 422 | 75 | |||
More Information
Set Membership: |
|

HDDC2
Genomic Map