Assay Details
| Gene Symbol: SCG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194420.2 | NP_001181349.2 | 103 | 69 | ||
| GenBank mRNA | JU336325.1 | - | 117 | 69 | ||
| JV703360.1 | - | 128 | 69 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194420.2 transcripts.
Set Membership: |
|

SCG2
Genomic Map