Assay Details
| Gene Symbol: PPM1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257613.2 | NP_001244542.1 | 352 | 68 | ||
| GenBank mRNA | JU335168.1 | - | 352 | 68 | ||
| JU488899.1 | - | 383 | 68 | |||
| JV047472.1 | - | 382 | 68 | |||
| JV717770.1 | - | 357 | 68 | |||
More Information
Set Membership: |
|

PPM1G
Genomic Map