Assay Details
| Gene Symbol: ATG4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261491.1 | NP_001248420.1 | 342 | 79 | ||
| XM_015127939.2 | XP_014983425.1 | 366 | 79 | |||
| XM_028841766.1 | XP_028697599.1 | 488 | 79 | |||
| GenBank mRNA | JU332815.1 | - | 344 | 79 | ||
| JU473196.1 | - | 342 | 79 | |||
More Information
Set Membership: |
|

ATG4A
Genomic Map