Assay Details
Gene Symbol: SIX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015143827.2 | XP_014999313.1 | 764 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
