Assay Details
| Gene Symbol: MOXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265792.1 | NP_001252721.1 | 1403 | 68 | ||
| GenBank mRNA | JU326645.1 | - | 1232 | 68 | ||
| JU326646.1 | - | 1232 | 68 | |||
| JV565025.1 | - | 1403 | 68 | |||
| JV711592.1 | - | 1300 | 68 | |||
More Information
Set Membership: |
|

MOXD1
Genomic Map