Assay Details
Gene Symbol: NR1H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194697.1 | NP_001181626.1 | 1028 | 71 | ||
XM_015124413.2 | XP_014979899.1 | 794 | 71 | |||
XM_015124414.2 | XP_014979900.1 | 974 | 71 | |||
XM_028838520.1 | XP_028694353.1 | 760 | 71 | |||
XM_028838521.1 | XP_028694354.1 | 503 | 71 | |||
GenBank mRNA | JU336224.1 | - | 965 | 71 | ||
JU472352.1 | - | 886 | 71 | |||
JV556720.1 | - | 976 | 71 | |||
JV627735.1 | - | 997 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
