Assay Details
| Gene Symbol: EFNA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265728.1 | NP_001252657.1 | 675 | 70 | ||
| XM_015140527.2 | XP_014996013.1 | 1151 | 70 | |||
| XM_028849187.1 | XP_028705020.1 | 1024 | 70 | |||
| GenBank mRNA | JU321748.1 | - | 453 | 70 | ||
| JV565076.1 | - | 675 | 70 | |||
| JV708763.1 | - | 652 | 70 | |||
More Information
Set Membership: |
|

EFNA5
Genomic Map