Assay Details
| Gene Symbol: TRNAU1AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133057.2 | XP_014988543.1 | 920 | 80 | ||
| XM_015133061.2 | XP_014988547.1 | 801 | 80 | |||
| XM_028837172.1 | XP_028693005.1 | 774 | 80 | |||
| GenBank mRNA | JU320938.1 | - | 793 | 80 | ||
| JU471912.1 | - | 803 | 80 | |||
| JV043489.1 | - | 731 | 80 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TRNAU1AP
Genomic Map