Assay Details
| Gene Symbol: CDIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141815.2 | XP_014997301.1 | 980 | 98 | ||
| XM_015141816.2 | XP_014997302.1 | 980 | 98 | |||
| XM_015141817.2 | XP_014997303.1 | 980 | 98 | |||
| XM_015141818.2 | XP_014997304.2 | 701 | 98 | |||
| XM_015141820.2 | XP_014997306.1 | 934 | 98 | |||
| XM_028850721.1 | XP_028706554.1 | 715 | 98 | |||
| XM_028850722.1 | XP_028706555.1 | 939 | 98 | |||
| GenBank mRNA | JU321014.1 | - | 961 | 98 | ||
| JU321187.1 | - | 961 | 98 | |||
| JU321192.1 | - | 961 | 98 | |||
| JU477516.1 | - | 851 | 98 | |||
More Information
Set Membership: |
|

CDIN1
Genomic Map