Assay Details
| Gene Symbol: CYLD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126108.2 | XP_014981594.1 | 1072 | 73 | ||
| XM_015126109.2 | XP_014981595.1 | 1105 | 73 | |||
| XM_015126111.2 | XP_014981597.1 | 1032 | 73 | |||
| XM_015126112.2 | XP_014981598.1 | 1072 | 73 | |||
| XM_015126113.2 | XP_014981599.1 | 1105 | 73 | |||
| GenBank mRNA | JU322238.1 | - | 1044 | 73 | ||
| JU333134.1 | - | 1044 | 73 | |||
| JU471648.1 | - | 1085 | 73 | |||
More Information
Set Membership: |
|

CYLD
Genomic Map