Assay Details
| Gene Symbol: AIF1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194341.2 | NP_001181270.1 | 348 | 73 | ||
| XM_015116531.2 | XP_014972017.1 | 288 | 73 | |||
| GenBank mRNA | JV704188.1 | - | 327 | 73 | ||
More Information
Set Membership: |
|

AIF1L
Genomic Map