Assay Details
| Gene Symbol: ABHD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261502.1 | NP_001248431.1 | 616 | 77 | ||
| GenBank mRNA | JU325762.1 | - | 544 | 77 | ||
| JU476281.1 | - | 597 | 77 | |||
| JV043244.1 | - | 616 | 77 | |||
| JV621363.1 | - | 637 | 77 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ABHD10
Genomic Map