Assay Details
| Gene Symbol: E2F6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266655.1 | NP_001253584.1 | 908 | 66 | ||
| XM_015111695.2 | XP_014967181.1 | 1076 | 66 | |||
| XM_015111696.2 | XP_014967182.1 | 888 | 66 | |||
| XM_015111697.2 | XP_014967183.1 | 867 | 66 | |||
| XM_028831445.1 | XP_028687278.1 | 797 | 66 | |||
| GenBank mRNA | JU326754.1 | - | 572 | 66 | ||
| JU474575.1 | - | 797 | 66 | |||
| JV046365.1 | - | 944 | 66 | |||
| JV635866.1 | - | 908 | 66 | |||
More Information
Set Membership: |
|

E2F6
Genomic Map