Assay Details
| Gene Symbol: EXOG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001086498.4 | XP_001086498.1 | 220 | 92 | ||
| GenBank mRNA | JV667067.1 | - | 116 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EXOG| Gene Symbol: EXOG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001086498.4 | XP_001086498.1 | 220 | 92 | ||
| GenBank mRNA | JV667067.1 | - | 116 | 92 | ||
Set Membership: |
|