Assay Details
| Gene Symbol: MPIG6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194580.2 | NP_001181509.1 | 630 | 85 | ||
| XM_015135936.2 | XP_014991422.1 | 951 | 85 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MPIG6B| Gene Symbol: MPIG6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194580.2 | NP_001181509.1 | 630 | 85 | ||
| XM_015135936.2 | XP_014991422.1 | 951 | 85 | |||
Set Membership: |
|