Assay Details
| Gene Symbol: BCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002798291.3 | XP_002798337.3 | 2570 | 70 | ||
| XM_028828640.1 | XP_028684473.1 | 2115 | 70 | |||
| XM_028828641.1 | XP_028684474.1 | 2115 | 70 | |||
| GenBank mRNA | JU327211.1 | - | 1695 | 70 | ||
| JU327212.1 | - | 1695 | 70 | |||
| JU491079.1 | - | 1694 | 70 | |||
| JV590013.1 | - | 1838 | 70 | |||
More Information
Set Membership: |
|

BCR
Genomic Map