Assay Details
| Gene Symbol: SPATA17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116149.2 | XP_014971635.1 | 581 | 76 | ||
| XM_028853294.1 | XP_028709127.1 | 530 | 76 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPATA17| Gene Symbol: SPATA17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116149.2 | XP_014971635.1 | 581 | 76 | ||
| XM_028853294.1 | XP_028709127.1 | 530 | 76 | |||
Set Membership: |
|