Assay Details
| Gene Symbol: RGS18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001113061.4 | XP_001113061.1 | 398 | 151 | ||
| GenBank mRNA | JV721149.1 | - | 328 | 151 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGS18| Gene Symbol: RGS18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001113061.4 | XP_001113061.1 | 398 | 151 | ||
| GenBank mRNA | JV721149.1 | - | 328 | 151 | ||
Set Membership: |
|