Assay Details
| Gene Symbol: RCSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125288.2 | XP_014980774.2 | 1642 | 73 | ||
| XM_015125296.2 | XP_014980782.2 | 591 | 73 | |||
| XM_028850088.1 | XP_028705921.1 | 554 | 73 | |||
| XM_028850097.1 | XP_028705930.1 | 1212 | 73 | |||
| GenBank mRNA | JU477831.1 | - | 653 | 73 | ||
| JU477832.1 | - | 563 | 73 | |||
More Information
Set Membership: |
|

RCSD1
Genomic Map