Assay Details
| Gene Symbol: OXTR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044732.1 | NP_001038197.1 | 926 | 86 | ||
| XM_028843186.1 | XP_028699019.1 | 1545 | 86 | |||
| GenBank mRNA | JV652705.1 | - | 169 | 86 | ||
| U82440.1 | - | 926 | 86 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

OXTR
Genomic Map