Assay Details
| Gene Symbol: LRP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260541.1 | NP_001247470.1 | 272 | 91 | ||
| XM_028838404.1 | XP_028694237.1 | 456 | 91 | |||
| XM_028838405.1 | XP_028694238.1 | 453 | 91 | |||
| GenBank mRNA | JU326046.1 | - | 253 | 91 | ||
| JV044861.1 | - | 272 | 91 | |||
| JV616677.1 | - | 397 | 91 | |||
| JV616678.1 | - | 397 | 91 | |||
| JV667249.1 | - | 290 | 91 | |||
| Gene Symbol: SLC7A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV714996.1 | - | 169 | 91 | ||
| JV714997.1 | - | 169 | 91 | |||
More Information
Set Membership: |
|

LRP3
Genomic Map