Assay Details
| Gene Symbol: FBXO32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257729.1 | NP_001244658.1 | 897 | 70 | ||
| GenBank mRNA | JU332809.1 | - | 897 | 70 | ||
| JV456031.1 | - | 74 | 70 | |||
More Information
Set Membership: |
|

FBXO32
Genomic Map