Assay Details
| Gene Symbol: RNF141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266485.1 | NP_001253414.1 | 309 | 61 | ||
| XM_015114744.2 | XP_014970230.1 | 429 | 61 | |||
| GenBank mRNA | JU319636.1 | - | 339 | 61 | ||
| JU319637.1 | - | 339 | 61 | |||
| JU332105.1 | - | 339 | 61 | |||
| JU332106.1 | - | 339 | 61 | |||
| JU332107.1 | - | 339 | 61 | |||
| JU332108.1 | - | 339 | 61 | |||
| JU473325.1 | - | 404 | 61 | |||
| JV044940.1 | - | 309 | 61 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RNF141
Genomic Map