Assay Details
| Gene Symbol: LOC705843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144470.2 | XP_014999956.1 | 441 | 67 | ||
| GenBank mRNA | JU327533.1 | - | 198 | 67 | ||
| JV531498.1 | - | 202 | 67 | |||
| JV667329.1 | - | 208 | 67 | |||
More Information
Set Membership: |
|

LOC705843
Genomic Map