Assay Details
Gene Symbol: DHRS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260537.2 | NP_001247466.1 | 653 | 79 | ||
GenBank mRNA | JU328603.1 | - | 429 | 79 | ||
JV044007.1 | - | 654 | 79 | |||
JV635508.1 | - | 649 | 79 | |||
JV669479.1 | - | 607 | 79 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
