Assay Details
| Gene Symbol: TOMM70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266153.1 | NP_001253082.1 | 802 | 65 | ||
| GenBank mRNA | JU322043.1 | - | 884 | 65 | ||
| JU470868.1 | - | 802 | 65 | |||
| JV527377.1 | - | 226 | 65 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TOMM70
Genomic Map