Assay Details
| Gene Symbol: ETV6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266080.1 | NP_001253009.1 | 1139 | 64 | ||
| XM_015151078.2 | XP_015006564.1 | 1127 | 64 | |||
| XM_028828809.1 | XP_028684642.1 | 3416 | 64 | |||
| GenBank mRNA | JU471226.1 | - | 1215 | 64 | ||
| JU471227.1 | - | 1215 | 64 | |||
| JV046436.1 | - | 1157 | 64 | |||
| JV635669.1 | - | 1139 | 64 | |||
More Information
Set Membership: |
|

ETV6
Genomic Map