Assay Details
| Gene Symbol: RAB35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194766.2 | NP_001181695.1 | 423 | 70 | ||
| XM_015152920.1 | XP_015008406.1 | 423 | 70 | |||
| GenBank mRNA | JU320504.1 | - | 357 | 70 | ||
| JU474110.1 | - | 400 | 70 | |||
| JV044838.1 | - | 428 | 70 | |||
| JV601370.1 | - | 410 | 70 | |||
More Information
Set Membership: |
|

RAB35
Genomic Map