Assay Details
Gene Symbol: LOC114675556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842777.1 | XP_028698610.1 | 2180 | 145 |
Gene Symbol: ACACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266778.1 | NP_001253707.1 | 3578 | 145 | ||
GenBank mRNA | JU320354.1 | - | 3527 | 145 | ||
JU322287.1 | - | 3578 | 145 | |||
JV565894.1 | - | 3610 | 145 |

More Information
Set Membership: |
![]() ![]() |
