Assay Details
| Gene Symbol: TXNDC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135522.2 | XP_014991008.1 | 764 | 65 | ||
| GenBank mRNA | JU325833.1 | - | 529 | 65 | ||
| JV697674.1 | - | 640 | 65 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TXNDC5
Genomic Map