Assay Details
| Gene Symbol: LRP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257931.1 | NP_001244860.1 | 1984 | 79 | ||
| GenBank mRNA | JU333904.1 | - | 1984 | 79 | ||
| JU333905.1 | - | 1984 | 79 | |||
| JU475093.1 | - | 1982 | 79 | |||
| JU475094.1 | - | 1982 | 79 | |||
| JV046703.1 | - | 1963 | 79 | |||
| JV046704.1 | - | 1963 | 79 | |||
| JV636091.1 | - | 1967 | 79 | |||
More Information
Set Membership: |
|

LRP10
Genomic Map