Assay Details
| Gene Symbol: MROH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193970.2 | NP_001180899.1 | 1004 | 95 | ||
| GenBank mRNA | JV571811.1 | - | 69 | 95 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MROH8| Gene Symbol: MROH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193970.2 | NP_001180899.1 | 1004 | 95 | ||
| GenBank mRNA | JV571811.1 | - | 69 | 95 | ||
Set Membership: |
|