Assay Details
| Gene Symbol: CNPY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194868.3 | NP_001181797.2 | 526 | 79 | ||
| XM_028846599.1 | XP_028702432.1 | 588 | 79 | |||
| GenBank mRNA | JU332581.1 | - | 200 | 79 | ||
| JU474048.1 | - | 295 | 79 | |||
| JV045968.1 | - | 295 | 79 | |||
| JV603863.1 | - | 328 | 79 | |||
| JV640514.1 | - | 336 | 79 | |||
More Information
Set Membership: |
|

CNPY3
Genomic Map