Assay Details
| Gene Symbol: WWOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105944.4 | XP_001105944.3 | 745 | 62 | ||
| XM_028841112.1 | XP_028696945.1 | 745 | 62 | |||
| XM_028841113.1 | XP_028696946.1 | 745 | 62 | |||
| GenBank mRNA | JU322737.1 | - | 214 | 62 | ||
| JU475976.1 | - | 125 | 62 | |||
| JV619171.1 | - | 203 | 62 | |||
More Information
Set Membership: |
|

WWOX
Genomic Map