Assay Details
| Gene Symbol: FOSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260911.1 | NP_001247840.1 | 775 | 83 | ||
| XM_015111925.2 | XP_014967411.1 | 1027 | 83 | |||
| GenBank mRNA | JV043790.1 | - | 775 | 83 | ||
| JV600895.1 | - | 1099 | 83 | |||
| JV638256.1 | - | 775 | 83 | |||
More Information
Set Membership: |
|

FOSL2
Genomic Map