Assay Details
| Gene Symbol: OGFRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136683.2 | XP_014992169.1 | 1285 | 91 | ||
| XM_015136684.2 | XP_014992170.1 | 1282 | 91 | |||
| XM_015136685.2 | XP_014992171.1 | 560 | 91 | |||
| GenBank mRNA | JU323158.1 | - | 759 | 91 | ||
| JV723371.1 | - | 811 | 91 | |||
More Information
Set Membership: |
|

OGFRL1
Genomic Map