Assay Details
| Gene Symbol: ADAMTS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116124.2 | XP_014971610.2 | 1658 | 138 | ||
| GenBank mRNA | JV569756.1 | - | 712 | 138 | ||
| JV705819.1 | - | 431 | 138 | |||
More Information
Set Membership: |
|

ADAMTS8
Genomic Map