Assay Details
| Gene Symbol: PGM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015138198.2 | XP_014993684.1 | 1088 | 67 | ||
| GenBank mRNA | JU322496.1 | - | 1021 | 67 | ||
| JU472974.1 | - | 1032 | 67 | |||
| JV565145.1 | - | 1024 | 67 | |||
| JV704179.1 | - | 1032 | 67 | |||
More Information
Set Membership: |
|

PGM2
Genomic Map