Assay Details
| Gene Symbol: PRDX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193811.2 | NP_001180740.1 | 588 | 84 | ||
| XM_015127083.2 | XP_014982569.1 | 615 | 84 | |||
| XM_015127085.2 | XP_014982571.1 | 693 | 84 | |||
| XM_028841690.1 | XP_028697523.1 | 615 | 84 | |||
| XM_028841691.1 | XP_028697524.1 | 615 | 84 | |||
| GenBank mRNA | JU321136.1 | - | 521 | 84 | ||
| JV045241.1 | - | 527 | 84 | |||
| JV566744.1 | - | 507 | 84 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PRDX4
Genomic Map