Assay Details
| Gene Symbol: PAX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015147955.2 | XP_015003441.2 | 300 | 61 | ||
| XM_015147956.2 | XP_015003442.2 | 305 | 61 | |||
| XM_015147957.2 | XP_015003443.2 | 305 | 61 | |||
| XM_015147958.2 | XP_015003444.2 | 300 | 61 | |||
| XM_015147959.2 | XP_015003445.2 | 298 | 61 | |||
| XM_015147960.2 | XP_015003446.2 | 302 | 61 | |||
| XM_015147961.2 | XP_015003447.2 | 298 | 61 | |||
| XM_028826770.1 | XP_028682603.1 | 306 | 61 | |||
More Information
Set Membership: |
|

PAX2
Genomic Map