Assay Details
| Gene Symbol: PRKAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266115.1 | NP_001253044.1 | 713 | 99 | ||
| XM_015152911.2 | XP_015008397.1 | 915 | 99 | |||
| XM_015152912.2 | XP_015008398.1 | 915 | 99 | |||
| XM_015152913.2 | XP_015008399.1 | 915 | 99 | |||
| GenBank mRNA | JU320569.1 | - | 689 | 99 | ||
| JU473536.1 | - | 717 | 99 | |||
| JV043683.1 | - | 713 | 99 | |||
More Information
Set Membership: |
|

PRKAB1
Genomic Map