Assay Details
| Gene Symbol: SLC39A13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266399.1 | NP_001253328.1 | 824 | 58 | ||
| XM_015114189.2 | XP_014969675.2 | 906 | 58 | |||
| XM_015114191.2 | XP_014969677.2 | 794 | 58 | |||
| XM_015114192.2 | XP_014969678.1 | 794 | 58 | |||
| XM_015114193.2 | XP_014969679.1 | 906 | 58 | |||
| XM_028832707.1 | XP_028688540.1 | 1225 | 58 | |||
| XM_028832708.1 | XP_028688541.1 | 1337 | 58 | |||
| XM_028832709.1 | XP_028688542.1 | 1243 | 58 | |||
| XM_028832710.1 | XP_028688543.1 | 1146 | 58 | |||
| GenBank mRNA | JU321503.1 | - | 824 | 58 | ||
| JV567244.1 | - | 798 | 58 | |||
More Information
Set Membership: |
|

SLC39A13
Genomic Map