Assay Details
| Gene Symbol: NRGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267599.3 | NP_001254528.1 | 48 | 55 | ||
| XM_015115926.2 | XP_014971412.1 | 222 | 55 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001267599.3, XM_015115926.2 transcripts.
Set Membership: |
|

NRGN
Genomic Map